Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Amphibian TGF-beta 5 Protein
Click to view available options
Quantity:
10 μg
2 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Description
The Recombinant Amphibian TGF-beta 5 Protein is derived from Sf 21 (baculovirus). The Recombinant Amphibian TGF-beta 5 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
| Gene ID (Entrez) | 397778 |
| Molecular Weight (g/mol) | 12.6 kDa |
| Name | TGF-beta 5 |
| Quantity | 2 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived amphibian TGF-beta 5 protein Gly271-Ser382 & Gln274-Ser382 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute at 20 μg/mL in sterile 4 mM HCl containing 0.1% human or bovine serum albumin, |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction