Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Bovine FGF basic/FGF2/bFGF Protein

Catalog No. 2099FB025
Click to view available options
Quantity:
25 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Bovine FGF basic/FGF2 Protein is derived from E. coli. The Recombinant Bovine FGF basic/FGF2 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number E. coli-derived bovine FGF basic/FGF2/bFGF protein Pro10-Ser155, with an N-terminal Ala
Formulation Lyophilized from a 0.2μm filtered solution in Tris-HCl with BSA as a carrier protein.
Gene ID (Entrez) 281161
Molecular Weight (g/mol) 16.5 kDa
Name FGF basic
Quantity 25 μg
Source E. coli-derived bovine FGF basic/FGF2 protein Pro10-Ser155, with an N-terminal Ala
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.