Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Bovine Fibronectin Protein

Catalog No. 1030FN01M
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to support cell attachment and spreading when used as a substratum for cell culture. In this application, the recommended concentration for this effect is 1-5 μg/cm2. Fibronectin can also be added in the media to support cell spreading at a concentration of 0.5-50 μg/mL. Optimal concentration depends on cell type as well as the application or research objectives. Measured by the ability of the immobilized protein to support the adhesion of B16-F1 mouse melanoma cells. The ED50 for this effect is 10.0-100 ng/mL.

Specifications

Formulation Supplied as a 0.2μm filtered solution in Tris-HCl, NaCl and Urea.
Gene ID (Entrez) 280794
Name Fibronectin
Quantity 1 mg
Source Bovine plasma-derived Fibronectin protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. Avoid vortexing and excessive agitation.
Conjugate Unconjugated
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.