Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Canine IFN-gamma ELISpot Kit

Catalog No. p-4765285
Click to view available options
Quantity:
1 x 96-well Plate
6 x 96-well Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

  • Microplate
  • Biotinylated Detection Antibody
  • Streptavidin conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • Positive Control

For the quantitative determination of the frequency of cells releasing canine IL-4.

Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells

Specifications

Antigen IFN-gamma
Cross Reactivity Dog
Includes Canine IFN-gamma Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Canine IFN-gamma Positive Control
Quantity 1 x 96-well Plate
Sample Type Whole Cells
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) Detection of cytokine secreting cells, quantitative sandwich ELISA
Product Type Canine IFN-gamma ELISpot Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.