Learn More
Description
- Fluoromethyl ketone (FMK)-derivatized peptides are useful tools for studying caspase activity
- Used in tissue cultures to inhibit apoptosis at final working concentrations of 50nM to 100μM
- Regulated by a variety of stimili, including APAF1, CFLAR/FLIP, NOL3/ARC, IAP, BIRC1/NAIP, BIRC2/cIAP-1, BIRC3/cIAP-2, BIRC4/XIAP, BIRC5/Survivin and BIRC7/Livin
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 1 mg |
| Product Type | Caspase-8 |
| Molecular Weight (g/mol) | 654Da |
| Product Line | R&D Systems |
| Inhibitors | Z-IETD-FMK |
| Form | Lyophilized |
For research use only. Not for use in diagnostic procedures
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

