Learn More
R&D Systems™ clasto-Lactacystin beta Lactone
Description
- The beta-lactone intermediate is generated from the aqueous hydrolysis of lactacystin and is the active inhibitory species that reacts with the proteasome
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 1 mg |
| Molecular Weight (g/mol) | 213.24 |
| Inhibitors | Proteasome |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.