Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CU-T12-9
GREENER_CHOICESDP

Catalog No. 541450 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
541450 50 mg
541410 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 541450 Supplier Tocris Bioscience™ Supplier No. 5414/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent TLR1/2 agonist

CU-T12-9 is a potent TLR1/2 agonist (EC50 = 52.9 nM). Activates the NF-κB pathway and upregulates proinflammatory cytokines in vitro. Also enhances TLR1 and TLR2 dimerization.

Specifications

Chemical Name or Material CU-T12-9
CAS 1821387-73-8
Synonym N-Methyl-4-nitro-2-(4-(4-trifluoromethyl)phenyl)-1H-imidazol-1-yl)aniline
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C17H13F3N4O2
Formula Weight 362.31
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >99%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.