Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Cultrex Poly-L-Lysine (2 x 100 mL)

Click to view available options
Quantity:
200 mL (2 x 100 mL)
Description
Applications: Substrate for cell culture adhesion. An area of 25 cm2 can be coated with 0.5 ml of a 0.01% Poly-L-Lysine solution. Optimal conditions for attachment must be determined for each cell line and application. Slides may be dipped in the solution and air dried before applying sample. Quality Control: Tested for ability to promote attachment of rat PC-12 pheochromocytoma cells. Cell Culture Qualified: No bacterial or fungal growth detected after incubation at 37degreesC for 14 days following USP XXIV, Chapter 71 sterility testing. No mycoplasma contamination detected by PCR.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Content And Storage | Product is stable for at least 6 months from the date of receipt when stored at 2 - 8° C. Keep sterile. |
Form | Liquid |
Product Type | Poly-L-Lysine |
Sterility | Sterile-filtered |
Endotoxin Level | <20.0 EU per 1μg of the protein by the LAL method |
Formulation | 0.01% in Phosphate Buffered Saline, sterile-filtered. |
For Use With (Application) | Bioactivity |
Quantity | 200 mL (2 x 100 mL) |
Regulatory Status | RUO |
Source | Fermentation using starch. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction