Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Edaglitazone
GREENER_CHOICE

Catalog No. 478410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
478410 10 mg
478450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 478410 Supplier Tocris Bioscience™ Supplier No. 4784/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective PPARγ agonist; antidiabetic

Edaglitazone is a potent and selective PPARγ agonist (EC50 values are 35.6 and 1053 nM for PPARγ and PPARα cofactor recruitment respectively. Enhances insulin sensitivity in obese, but not lean, rats. Antidiabetic and orally bioavailable.

Specifications

Chemical Name or Material Edaglitazone
CAS 213411-83-7
Synonym 5-[[4-[2-(5-Methyl-2-phenyl-4-oxazolyl)ethoxy]benzo[b]thien-7-yl]methyl]-2,4-thiazolidinedione
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C24H20N2O4S2
Formula Weight 464.56
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.