Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ FICZ
GREENER_CHOICESDP

Catalog No. 53041 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53041 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 53041 Supplier R&D Systems™ Supplier No. 5304/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. High affinity aryl hydrocarbon receptor (AhR) agonist

FICZ is a high affinity aryl hydrocarbon receptor (AhR) agonist (Kd = 70 pM). Proposed to be an endogenous AhR ligand. Induces transient expression of cytochrome P450-1A1 (CYP1A1) in vitro.

Specifications

Chemical Name or Material FICZ
CAS 172922-91-7
Synonym 5,11-Dihydroindolo[3,2-b]carbazole-6-carboxaldehyde
Quantity 1 mg
Solubility Soluble to 25mM in DMSO
Product Line Tocris
Molecular Formula C19H12N2O
Formula Weight 284.31
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >95%
Show More Show Less

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.