Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flow Cytometry Fixation/Permeabilization Buffer I (1X), R&D Systems™

Catalog No. fc007
Click to view available options
Quantity:
125mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Applications: Flow Cytometry

  • Designed for intracellular flow cytometry applications with nonconjugated or fluorochrome-conjugated antibodies

Applications:

Flow Cytometry

Specifications

Product Type Fixation/Permeabilization Buffer
Content And Storage Store the unopened product at 2°C to 8°C for up to 6 months from date of receipt.
Stability Upto 6 Months
Formulation Sodium azide 0.05% and formaldehyde 4%
Quantity 125mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.