Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flow Cytometry Fixation and Permeabilization Buffer Kit I, R&D Systems™
GSA_VA

Catalog No. fc009
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Applications: Flow Cytometry

  • Designed for intracellular flow cytometry applications with nonconjugated or fluorochrome-conjugated antibodies
  • Includes 100mL of Flow Cytometry Fixation Buffer (1X) and 250mL of Flow Cytometry Permeabilization/Wash Buffer I (1X)

Applications:

Flow Cytometry

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 1 Kit
Product Type Fixation/Permeabilization Buffer
Stability Upto 6 Months
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Formulation Sodium azide 0.05% and formaldehyde 4%
Includes 100mL flow cytometry fixation buffer 1X, 250mL flow cytometry permeabilization/wash buffer I 1X
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.