Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flow Cytometry Human Lyse Buffer (10X), R&D Systems™

Catalog No. fc002
Click to view available options
Quantity:
50mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Applications: Flow Cytometry

  • Optimized to prepare human whole blood samples or other cell preparations containing an excess of red blood cells

Applications:

Flow Cytometry

Specifications

Product Type Lysis Buffer
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Formulation Human RBC's
Quantity 50mL
Stability Upto 6 Months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.