Description
Conjugation of the protein of interest, if already contained in the HeLa fraction, may be tracked via immuno-detection, or if added exogenously, by immunodetection or radiolabel
Specifications
Specifications
| Description | Fraction II Conjugation Kit, Rabbit Reticulocyte |
| For Use With (Application) | Controlled conjugation of exogenously added or endogenously contained substrate proteins through the ubiquitin proteasome pathway (UPP) |
| Quantity | 1 Kit |
| Product Line | R&D Systems |