Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GlyH 101
GREENER_CHOICE

Catalog No. 548510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
548510 10 mg
548550 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 548510 Supplier Tocris Bioscience™ Supplier No. 5485/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reversible, voltage-dependent CFTR chloride channel blocker

GlyH 101 is a reversible, voltage-dependent CFTR chloride channel blocker (Ki = 4.3 μM). Inhibits forskolin -induced hyperpolarization in nasal potential differences and inhibits cholera toxin-induced intestinal fluid secretion in mice.

Specifications

Chemical Name or Material GlyH 101
CAS 328541-79-3
Synonym N-2-Naphthalenyl-2-[(3,5-dibromo-2,4-dihydroxyphenyl)methylene]glycine hydrazide
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C19H15Br2N3O3
Formula Weight 493.15
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.