Learn More
R&D Systems™ HT Superoxide Dismutase Assay Kit
SDP
Catalog No. 7501500K Shop All Novus Products
Change view
Click to view available options
Quantity:
480 Tests
Catalog No. Quantity
7501500K 480 Tests
1 options
This item is not returnable. View return policy
Catalog No. 7501500K Supplier R&D Systems™ Supplier No. 7501500K
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
SOD, SOD Reaction Buffer, Xanthine Solution, Xanthine Oxidase Solution, Triton™ X-100, WST-1 Reagent, Five 96-well plates for 480 tests
A 96-well microplate-based assay designed for high-throughput analysis of Superoxide Dismutase (SOD) activity in mammalian tissue and cell lysates
SODs are metalloenzymes that catalyze the dismutation of the superoxide radical into hydrogen peroxide and molecular oxygen, providing an important defense against oxidative damage.
Free of interference by other catalytic activities and well-suited to assaying SOD in mammalian cell lysates
Not greatly disturbed by trace metals
Performed in as little as 6 minutes
The superoxide anion then coverts the tetrazolium salt WST-1 to the c
Specifications
Content And Storage The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Product Line R&D Systems
Quantity 480 Tests