Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Active Coagulation Factor XIV/ProteinC

Catalog No. 4998SE010
Click to view available options
:
10ug; Carrier Free
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Active Coagulation Factor XIV/ProteinC is derived from CHO. The Recombinant Human Active Coagulation Factor XIV/ProteinC has been validated for the following applications: Enzyme Activity.

Specifications

Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 .
Gene ID (Entrez) 5624
Molecular Weight (g/mol) 19 kDa (light chain), 29 kDa (heavy chain)
Name Active Coagulation Factor XIV/Protein C
Quantity 10μg
Source Chinese Hamster Ovary cell line,CHO-derived human Coagulation Factor XIV/Protein C protein Ala43-Pro461,with a C-terminal 10-His tag The isolated protein was activated by thrombin/thrombomodulin complex and further purified.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Structural Form Disulfide-linked heterodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.