Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human ADAM10 Protein
Click to view available options
Quantity:
20 μg
Description
The Recombinant Human ADAM10 Protein is derived from Sf 21 (baculovirus). The Recombinant Human ADAM10 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Formulation | Supplied as a 0.2 μm filtered solution in MES, NaCl, ZnCl 2Glycerol and Brij-35. |
Gene ID (Entrez) | 102 |
Molecular Weight (g/mol) | 52 kDa |
Name | ADAM10 |
Quantity | 20 μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human ADAM10 protein Thr214-Glu672,with a C-terminal 10-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
Structural Form | Recombinant Human ADAM10 is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction