Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human ADAM8 Protein
Click to view available options
Quantity:
20 μg
Description
The Recombinant Human ADAM8 Protein is derived from NS0. The Recombinant Human ADAM8 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Formulation | Supplied as a 0.2 μm filtered solution in Glycerol, Tris, NaCl and CaCl 2 . |
Gene ID (Entrez) | 101 |
Molecular Weight (g/mol) | 38 kDa |
Name | ADAM8 aa 158-497 |
Quantity | 20 μg |
Source | Mouse myeloma cell line,NS0-derived human ADAM8 protein Met1-Pro497,with a C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
Structural Form | Recombinant Human ADAM8 is prone to proteolytic cleavage at C-terminus.The poly-Histag may not be present in the preparation. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction