Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Arginase 1/ARG1 Protein

Catalog No. 5868AR010
Click to view available options
Quantity:
10 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by the production of urea during the hydrolysis of arginine. The specific activity is >35,000 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P05089
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, Glycerol and DTT.
Gene ID (Entrez) 383
Molecular Weight (g/mol) M.W. (Observed): 40 kDa, reducing conditions; M.W. (theoretical): 36 kDa
Name Arginase 1/ARG1
Quantity 10 μg
Source E. coli-derived human Arginase 1/ARG1 protein Met1-Lys322 with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Structural Form Monomer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol ARG1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Arginase 1/ARG1/liver Arginase
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.