Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human B7-H3 (4Ig)/B7-H3b Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Description
Measured by its ability to inhibit anti-CD3-induced proliferation of stimulated human T cells. T lymphocytes cultured for 72 hours with PHA were incubated for an additional 3 days in 96 well plates coated with 500 ng/mL anti-CD3 and/or 10 μg/mL rhB7-H3 (4Ig)/B7-H3b. The presence of rhB7-H3 (4Ig)/B7-H3b at 10 μg/mL inhibited the anti-CD3 response by >30%. Optimal dilutions should be determined by each laboratory for each application.
Specifications
Specifications
| Accession Number | Q5ZPR3 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 80381 |
| Molecular Weight (g/mol) | M.W. (Observed): 78-90 kDa, reducing conditions; M.W. (theoretical): 48 kDa |
| Name | B7-H3 (4Ig)/B7-H3b |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived human B7-H3 protein Gly27-Thr461, with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction