Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human beta-NGF Protein
GSA_VA

Catalog No. p-4719138
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetic Acid with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetic Acid.
Molecular Weight (g/mol):
13.5 kDa
13.5 kDa (monomer)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation Molecular Weight (g/mol)
256GF100 Lyophilized from a 0.2μm filtered solution in Acetic Acid with BSA as a carrier protein. 13.5 kDa (monomer)
256GF100CF Lyophilized from a 0.2μm filtered solution in Acetic Acid. 13.5 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 256GF100 Supplier R&D Systems™ Supplier No. 256GF100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human beta-NGF Protein is derived from NS0. The Recombinant Human beta-NGF Protein has been validated for the following applications: Bioactivity.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetic Acid with BSA as a carrier protein.
Gene ID (Entrez) 4803
Molecular Weight (g/mol) 13.5 kDa (monomer)
Name beta-NGF
Quantity 100 μg
Source Mouse myeloma cell line,NS0-derived human beta-NGF protein Ser122-Ala241
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.