Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
R&D Systems™ Recombinant Human Biotinidase/BTD Protein
                    Click to view available options
                
                
                    
                            
                                :
                            
                        
                        
                            
                                10ug; Carrier Free
                            
                        
                    Description
The Recombinant Human Biotinidase/BTD Protein is derived from CHO. The Recombinant Human Biotinidase/BTD Protein has been validated for the following applications: Enzyme Activity.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
                
            Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in NaH 2 PO 4 NaCl, EDTA and DTT. | 
| Gene ID (Entrez) | 686 | 
| Molecular Weight (g/mol) | 58 kDa | 
| Name | Biotinidase/BTD | 
| Quantity | 10μg | 
| Source | Chinese Hamster Ovary cell line,CHO-derived human Biotinidase/BTD protein Ala42-Asp543,with a C-terminal 6-His tag | 
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | 
| Conjugate | Unconjugated | 
| Recombinant | Recombinant | 
| Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie Blue stain at 5 μg per lane. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            