Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-15 Protein
Click to view available options
:
5ug
Description
The Recombinant Human BMP-15 Protein is derived from CHO. The Recombinant Human BMP-15 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
Gene ID (Entrez) | 9210 |
Molecular Weight (g/mol) | 13.9 kDa |
Name | BMP-15/GDF-9B |
Quantity | 5μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human BMP-15/GDF-9B protein Gln268-Arg392 (Thr266Arg) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Gene Alias | BMP15, BMP-15, bone morphogenetic protein 15, GDF-9 B, Growth/differentiation factor 9 B, ODG2, POF4 |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction