Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-9 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in Acetonitrile and TFA.
Description
The Recombinant Human BMP-9 Protein is derived from CHO. The Recombinant Human BMP-9 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Supplied as a 0.2μm filtered solution in Acetonitrile and TFA. |
Gene ID (Entrez) | 2658 |
Molecular Weight (g/mol) | 12.1 kDa (monomer) |
Name | BMP-9 |
Quantity | 10 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human BMP-9 protein Ser320-Arg429 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
Structural Form | Disulfide-linked homodimer |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction