Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CHST2 Protein

Catalog No. 5107st020
Click to view available options
:
20ug; Carrier Free
This item is not returnable. View return policy

Catalog No. 5107ST020

Supplier: R&D Systems™ 5107ST020

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to transfer sulfate from PAPS to N-acetyl-D-glucosamine. The specific activity is >125 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q9Y4C5
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 9435
Molecular Weight (g/mol) M.W. (Observed): 50-60 kDa, reducing conditions; M.W. (theoretical): 50 kDa
Name Carbohydrate Sulfotransferase 2/CHST2
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Carbohydrate Sulfotransferase 2/CHST2 protein Tyr76-Leu530, with an N-terminal 5-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol CHST2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Carbohydrate Sulfotransferase 2/CHST2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.