Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Carboxylesterase 1/CES1 Protein
Click to view available options
:
10ug; Carrier Free
Description
Measured by its ability to hydrolyze p-nitrophenylacetate. The specific activity is >6,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
Accession Number | NP_001020365 |
For Use With (Application) | Enzyme Activity |
Formulation | Supplied as a 0.2 μm filtered solution in Sodium Acetate and NaCl. |
Gene ID (Entrez) | 1066 |
Molecular Weight (g/mol) | M.W. (Observed): 61 kDa, reducing conditions; M.W. (theoretical): 61 kDa |
Name | Carboxylesterase 1/CES1 |
Quantity | 10 μg |
Source | Mouse myeloma cell line, NS0-derived human Carboxylesterase 1/CES1 protein His19-Glu563, with a C-terminal 10-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction