Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CCL5/RANTES DuoSet ELISA

Catalog No. dy27805
Click to view available options
Quantity:
5 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Human RANTES Capture Antibody; Human RANTES Detection Antibody; Human RANTES Standard; Streptavidin-HRP

Human CCL5/RANTES DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CCL5/RANTES DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CCL5/RANTES.

Specifications

Conjugate Biotin
Target Species Human
For Use With (Application) ELISA
Quantity 5 Plates
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.