Learn More
Description
The T cell receptor (TCR)/CD3 complex consists of two variable antigen-recognition receptor chains (TCR-alpha/TCR-beta or TCR-gamma/TCR-delta) that are non-covalently linked to at least four different invariant chains: CD3 gamma, CD3 delta, CD3 epsilon, and the CD3 zeta chain that exists as either a homodimer or as a heterodimer with its splice-variant the CD3 eta chain. The CD3 complex functions to transduce intracellular signals during TCR antigen recognition. CD3 is expressed early in thymocyte development, and on a subset of NK cells. CD3 epsilon is one of at least three invariant proteins that associate with the variable antigen recognition chains of the T cell receptor and function in signal transduction.
The resulting column eluate is a highly enriched T cell population with minimal depletion of T cell constituents
Total CD3+ cell recovery from CD3+ cells loaded ranges between 32% and 40%
Purity of recovered cells ranges between 85% and 95%
These enriched T cell populations are then available for tissue culture, activation studies, tissue typing, immune status monitoring, and flow cytometry
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| For Use With (Application) | Designed to prepare purified human T cell populations via high affinity negative selection |
| Includes | Human T Cell Selection Columns; 10X Column Buffer Concentrate |
| Product Line | R&D Systems |
| Quantity | 25/Pk. |
| Type | Human CD3+ T Cell Enrichment Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
