Learn More
Description
CD3+ is a universal marker for T cells. Enriched T cell populations are then available for cell culture, activation studies, phenotyping, immune status monitoring, and flow cytometry.
Total CD3+ cell recovery ranges between 30% and 50% of initial CD3+ cells loaded
The purity of recovered cells ranges from 85% to 95%
Based upon gravitational flow
Simple procedure
No specialized equipment required
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| For Use With (Application) | Designed to prepare purified human T cell populations via high affinity negative selection |
| Includes | Human T Cell Selection Columns; 10X Column Buffer Concentrate |
| Product Line | R&D Systems |
| Quantity | 5/Pk. |
| Type | Human CD3+ T Cell Enrichment Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
