Learn More
Description
The resulting column eluate is a highly enriched T cell subset population with minimal contamination of either CD45RA+ or CD8+ cells
Virtually no B cells, monocytes, or nonselected T cell subsets
Recovery of CD4+/45RA- cells from total CD4+/45RA- cells ranged between 8% and 25%
Purity (CD4+/45RA-) of recovered cells ranged between 95% and 99%, with no detectable CD4+/45RA+ or CD3+/8+ cells among recovered cells
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 4/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
