Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human cIAP-1 (HIAP-2) Protein

Catalog No. 818IA050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity

Measured by its ability to inhibit DEVD-AFC cleavage activity in cell extracts activated by addition of cytochrome c and dATP. The IC50 for this effect is 25-1000 nM. Optimal dilutions should be determined by each laboratory for each application.

Specifications

Accession Number Q13490
For Use With (Application) Inhibition Assay
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 329
Molecular Weight (g/mol) M.W. (Observed): 68 kDa, reducing conditions; M.W. (theoretical): 72 kDa
Name cIAP-1/HIAP-2
Quantity 50 μg
Source E. coli-derived human cIAP-1/HIAP-2 protein ATVID 10-His tag SIEGRA Human cIAP-1 (His2-Ser618) (N-terminus) Accession # Q13490 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol BIRC2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein cIAP-1/HIAP-2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.