Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Complement Component C1r Protein
Click to view available options
Quantity:
10 μg
Description
The Recombinant Human Complement Component C1r Protein is derived from NS0. The Recombinant Human Complement Component C1r Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in Tris and NaCl. |
Gene ID (Entrez) | 715 |
Molecular Weight (g/mol) | 51 kDa (A chain) & 28 kDa (B chain) |
Name | Complement Component C1r |
Quantity | 10 μg |
Source | Mouse myeloma cell line,NS0-derived human Complement Component C1r protein Met1-Asp705,with a C-terminal 10-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked heterodimer |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction