Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Complement Component C1s Protein
GSA_VA

Catalog No. 2060se010
Click to view available options
Quantity:
10 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Complement Component C1s Protein is derived from NS0. The Recombinant Human Complement Component C1s Protein has been validated for the following applications: Enzyme Activity.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl 2 and Glycerol.
Gene ID (Entrez) 716
Molecular Weight (g/mol) 47 kDa (A chain) & 28 kDa (B chain)
Name Complement Component C1s
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived human Complement Component C1s protein Met1-Asp688,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Structural Form Disulfide-linked heterodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.