Learn More
R&D Systems™ Human Deconjugating Enzyme Kits & Reagents Protein Sampler Pack
Shop All Novus Products

Description
Regulatory Status: RUO
Specifications
Specifications
| Host Species | Human |
| Content And Storage | Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Conjugate | Unconjugated |
| Quantity | 1 Kit |
| Product Type | Deconjugating Enzyme Kits and Reagents |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.