Learn More
Description
Regulatory Status: RUO
Specifications
Specifications
| Host Species | Human |
| Conjugate | Unconjugated |
| Quantity | 1 Kit |
| Product Type | Deconjugating Enzyme Kits and Reagents FRET K48-linked Substrate Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
