Learn More
R&D Systems™ Human Pluripotent Stem Cell Marker Antibody Panel
Click to view available options
Quantity:
1 Kit
Description
Includes
Mouse anti-alkaline phosphatase, Goat anti-Nanog, Goat anti-Oct-3/4, Mouse anti-SSEA-1, Mouse anti-SSEA-4
A cost-effective selection of five pluripotency marker antibodies: anti-alkaline phosphatase, anti-Nanog, anti-Oct-3/4, anti-SSEA-1, and anti-SSEA-4
Verifies the pluripotency of starting stem cell population with straightforward protocol
Specifications
Specifications
Quantity | 1 Kit |
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |