Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Erythropoietin/EPO (Ultrapure) Protein, CF

Catalog No. 286ep250
Click to view available options
Quantity:
250 U
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Erythropoietin (Ultrapure) Protein is derived from CHO. The Recombinant Human Erythropoietin (Ultrapure) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P01588
Formulation Supplied as a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 2056
Molecular Weight (g/mol) 21 kDa
Name Erythropoietin/EPO
Quantity 250 U
Source Chinese Hamster Ovary cell line, CHO-derived human Erythropoietin/EPO protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening.
Gene Alias ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.