Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FGF-5 Protein

Catalog No. 237F5050CF
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with Trehalose and with BSA as a carrier protein.
Supplied as a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT.
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human FGF-5 Protein is derived from E. coli. The Recombinant Human FGF-5 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Supplied as a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT.
Gene ID (Entrez) 2250
Molecular Weight (g/mol) 27 kDa
Name FGF-5
Quantity 50 μg
Source E. coli-derived human FGF-5 protein Glu23-Gly268 (Lys238Asn and Pro245Ser),with an N-terminal Met & Leu26-Gly268 (Lys238Asn and Pro245Ser)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.