Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human FGF basic/FGF2/bFGF (157 aa) Protein
Click to view available options
Quantity:
25 μg
Description
The Recombinant Human FGF basic/FGF2 (157 aa) Protein is derived from E. coli. The Recombinant Human FGF basic/FGF2 (157 aa) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris-HCl and NaCl. |
| Gene ID (Entrez) | 2247 |
| Molecular Weight (g/mol) | 17.4 kDa |
| Name | FGF basic 157 aa |
| Quantity | 25 μg |
| Source | E. coli-derived human FGF basic/FGF2/bFGF protein Gly132-Ser288 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Gene Alias | Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction