Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human GFR alpha-3 Fc Chimera (CHO-expressed)
Click to view available options
Quantity:
100 μg
Description
The Recombinant Human GFR alpha-3 Fc Chimera (CHO-expressed) is derived from CHO. The Recombinant Human GFR alpha-3 Fc Chimera (CHO-expressed) has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 2676 |
Molecular Weight (g/mol) | 64.8 kDa (monomer) |
Name | GFR alpha-3/GDNF R alpha-3 Fc Chimera |
Quantity | 100 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human GFR alpha-3/GDNF R alpha-3 protein, (N-terminus) Human GFR alpha-3 (Asp32-Asn374) Accession &Num; O60609 IEGRD Human IgG1 (Pro100-Lys330) (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction