Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human His6 Ubiquitin Mutant No K Recombinant Protein
Click to view available options
:
1mg; Carrier Free
Description
The Recombinant Human His6-Ubiquitin Mutant No K Protein is derived from E. coli. The Recombinant Human His6-Ubiquitin Mutant No K Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Formulation | Lyophilized from a solution in deionized water. |
Gene ID (Entrez) | 7314 |
Molecular Weight (g/mol) | 9.8 kDa |
Name | His6 Ubiquitin Mutant No K |
Quantity | 1mg |
Source | E. coli-derived human Ubiquitin protein Contains a 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction