Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Hyaluronidase 1/HYAL1 Protein

Catalog No. 7358GH020
Click to view available options
:
20ug; Carrier Free
This item is not returnable. View return policy

Catalog No. 7358GH020

Supplier: R&D Systems™ 7358GH020

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to hydrolyze S-35 labeled hyaluronan. <20 ng of rhHYAL1 is required for the 50% hydrolysis of 10 μg hyaluronan, as measured under the described conditions.

Specifications

Accession Number Q12794
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 3373
Molecular Weight (g/mol) M.W. (Observed): 50-60 kDa, reducing conditions; M.W. (theoretical): 47 kDa
Name Hyaluronidase 1/HYAL1
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Hyaluronidase 1/HYAL1 protein Met1-Trp435, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 3.2.1.35, FUS2, HYAL1, HYAL-1, hyaluronidase 1, hyaluronoglucosaminidase 1, Hyaluronoglucosaminidase-1, luCa-1, LUCA1, LUCA1hyaluronidase-1, Lung carcinoma protein 1, MGC45987, NAT6, plasma hyaluronidase, tumor suppressor LUCA-1
Gene Symbol HYAL1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein Hyaluronidase 1/HYAL1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.