Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IL-15R alpha Fc Chimera Protein
Click to view available options
Quantity:
100 μg
Description
The Recombinant Human IL-15R alpha Fc Chimera Protein is derived from Sf 21 (baculovirus). The Recombinant Human IL-15R alpha Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 3601 |
Molecular Weight (g/mol) | 42.6 kDa (monomer) |
Name | IL-15R alpha |
Quantity | 100 μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-15 R alpha protein, (N-terminus) Human IL-15 R alpha (Ile31-Thr172) Accession &Num; EAW86418 IEGRDMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
Gene Alias | CD215, CD215 antigen, IL15 R alpha, IL-15 R alpha, IL-15 receptor subunit alpha, IL15RA, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction