Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IL-17B (CHO) Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Sodium Phosphate and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Sodium Phosphate and NaCl.
Description
The Recombinant Human IL-17B (CHO) Protein is derived from CHO. The Recombinant Human IL-17B (CHO) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Sodium Phosphate and NaCl with BSA as a carrier protein. |
| Gene ID (Entrez) | 27190 |
| Molecular Weight (g/mol) | 18 kDa (monomer) |
| Name | IL-17B |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human IL-17B protein Gln21-Phe180 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction