Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human IL-2 ELISpot Kit
Click to view available options
Quantity:
1 x 96-well Plate
6 x 96-well Plates
Description
Includes
- Microplate
- Biotinylated Detection Antibody
- Streptavidin conjugated to Alkaline Phosphatase
- Dilution Buffers
- Wash Buffer Concentrate
- BCIP/NBT Chromogen
- Positive Control
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously.
Product Features:
- Detect and quantitate individual cells secreting
- High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
- No in vitro expansion of cells required
- High-throughput - ELISpot assays use only a small number of primary cells
Specifications
Specifications
Includes | One 96-well PVDF-backed plate coated w/polyclonal Ab for human IL-2, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT chromogen, Positive control |
Quantity | 1 x 96-well Plate |
Sample Type | Whole Cells |
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
For Use With (Application) | Diagnostics |
Product Type | Human IL-2 ELISpot Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction