Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IL-27 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HEPES, NaCl, EDTA and CHAPS with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in HEPES, NaCl, EDTA and CHAPS.
Description
The Recombinant Human IL-27 Protein is derived from NS0. The Recombinant Human IL-27 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Supplied as a 0.2μm filtered solution in HEPES, NaCl, EDTA and CHAPS. |
Gene ID (Entrez) | 246778 |
Molecular Weight (g/mol) | 50 kDa |
Name | IL-27 |
Quantity | 10 μg |
Source | Mouse myeloma cell line,NS0-derived human IL-27 protein, (N-terminus) Human IL-27 EBI-3 (Arg21-Lys229) Accession &Num; Q14213.2 GSGSSRGGSGSGGSGGGGSKL Human IL-27 p28 (Phe29-Pro243) Accession &Num; AAM34498 6-His tag (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction