Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IL-3 Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2um filtered solution in PBS with BSA as a carrier protein.
Description
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.02-0.1 ng/mL. The specific activity of Recombinant Human IL-3 is approximately 1.8 x 103 IU/μg, which is calibrated against recombinant human IL-3 WHO International Standard (NIBSC code: 91/510). Specific activity is for reference purposes only and is not routinely tested.
Specifications
Specifications
Accession Number | AAC08706 |
Formulation | Lyophilized from a 0.2um filtered solution in PBS with BSA as a carrier protein. |
Gene ID (Entrez) | 3562 |
Molecular Weight (g/mol) | 15 kDa |
Name | IL-3 |
Quantity | 250 μg |
Source | E. coli-derived human IL-3 protein Ala20-Phe152,with and without an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Gene Alias | Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction