Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Integrin alpha 2 beta 1 Protein

Catalog No. 5698a2050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. When bovine collagen-II is coated at 10 μg/mL, Recombinant Human Integrin alpha 2 beta 1 binds with an apparent Kd <1nM.

Specifications

Accession Number NP_002194 (Integrin alpha 2) & P05556 (Integrin beta 1)
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3673
Molecular Weight (g/mol) M.W. (Observed): 140-150 kDa (alpha 2) & 120-130 kDa (beta 1), reducing conditions; M.W. (theoretical): 126 kDa (alpha 2) & 83.3 kDa (beta 1)
Name Integrin alpha 2 beta 1
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Integrin alpha 2 beta 1 protein Human Integrin alpha 2 (Tyr30-Thr1132) (N-terminus) Accession # NP_002194 HPGGGSGGGS Human c-Jun (Arg276-His315) Human Integrin beta 1 (Gln21-Asp728) (N-terminus) Accession # P05556 GGGSGGGS Human c-Fos (Leu161-Ala199) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol ITGA2
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Integrin alpha 2 beta 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.