Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human KGF/FGF-7 Protein

Catalog No. p-4720172
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA.
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using 4MBr-5 rhesus monkey epithelial cells. Rubin, J.S. et al. (1989) Proc. Natl. Acad. Sci. USA 86:802. The ED50 for this effect is typically 6-60 ng/mL.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA with BSA as a carrier protein.
Gene ID (Entrez) 2252
Molecular Weight (g/mol) 19 kDa
Name KGF/FGF-7
Quantity 10 μg
Source E. coli-derived human KGF/FGF-7 protein Cys32-Thr194,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor)
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.